EMAIL THIS PAGE TO A FRIEND
Molecular Carcinogenesis
Zebularine inhibits the growth of A549 lung cancer cells via cell cycle arrest and apoptosis.
PMID 23661569
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PRS3805
Anti-AIM (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS3807
Anti-AIM (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS3335
Anti-Bcl-2 (ab1) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
PRS3337
Anti-Bcl-2 (ab2) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
B9804 Anti-Bcl-2 antibody, Mouse monoclonal, clone 10C4, purified from hybridoma cell culture
SAB4300339
Anti-BCL2 (Ab-56) antibody produced in rabbit, affinity isolated antibody
SAB4300340
Anti-BCL2 (Ab-70) antibody produced in rabbit, affinity isolated antibody
SAB2500154
Anti-BCL2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
AV01005
Anti-BCL2 antibody produced in rabbit, affinity isolated antibody
GW22659 Anti-DNMT1 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2500321
Anti-DNMT1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
D4692
Anti-DNMT1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
D4567
Anti-DNMT1 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
HPA002694
Anti-DNMT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2106406
Anti-DNMT1 antibody produced in rabbit, affinity isolated antibody
SAB1306667
ANTI-P53 (S15) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1306713
ANTI-P53 (S315) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1306623
ANTI-P53 (SUMOYLATION SITE SPECIFIC) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
P4235 Anti-p53 antibody produced in sheep, fractionated antiserum, buffered aqueous solution
SAB4300365
Anti-TP53 (Ab-18) antibody produced in rabbit, affinity isolated antibody
SAB4300369
Anti-TP53 (Ab-315) antibody produced in rabbit, affinity isolated antibody
SAB4300366
Anti-TP53 (Ab-33) antibody produced in rabbit, affinity isolated antibody
SAB4300367
Anti-TP53 (Ab-37) antibody produced in rabbit, affinity isolated antibody
SAB4300368
Anti-TP53 (Ab-46) antibody produced in rabbit, affinity isolated antibody
SAB4300362
Anti-TP53 (Ab-6) antibody produced in rabbit, affinity isolated antibody
SAB4300363
Anti-TP53 (Ab-9) antibody produced in rabbit, affinity isolated antibody
SAB2102511
Anti-TP53 (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2102512
Anti-TP53 (ab2) antibody produced in rabbit, affinity isolated antibody
SAB2102513
Anti-TP53 (ab3) antibody produced in rabbit, affinity isolated antibody
SAB2102514
Anti-TP53 (ab4) antibody produced in rabbit, affinity isolated antibody