EMAIL THIS PAGE TO A FRIEND
Zhurnal evoliutsionnoi biokhimii i fiziologii
[Effect of population density on enzymatic activity of antioxidative and phenol oxidase systems of imagoes and nymphs of the marble cockroach Nauphoeta cinerea].
PMID 23662478
Abstract
The work deals with effect of density of population on functional activity of components pf protective system of adult individuals and nymphs of the marble cockroach. The resistance of individuals has been noted to decrease both at individual maintenance and under conditions of overpopulation. Changes in activities of enzymes of antioxidative and phenoloxidase systems are studied ion the insect hemolymph and intestine. Possible consequences of isolation and overpopulation are discussed both for stability and for individual development.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C3515 Catalase from Aspergillus niger, ammonium sulfate suspension, ≥4,000 units/mg protein
C1345 Catalase from bovine liver, powder, suitable for cell culture, 2,000-5,000 units/mg protein
C9322 Catalase from bovine liver, lyophilized powder, 2,000-5,000 units/mg protein
C30 Catalase from bovine liver, aqueous suspension, 10,000-40,000 units/mg protein
C40 Catalase from bovine liver, lyophilized powder, ≥10,000 units/mg protein
C3155 Catalase from bovine liver, aqueous solution, ≥30,000 units/mg protein
C100 Catalase from bovine liver, aqueous suspension, 40,000-60,000 units/mg protein (E1%/405)
02071 Catalase from Corynebacterium glutamicum, solution, deep brown, ≥500000 U/mL
C3556 Catalase from human erythrocytes, ≥90% (SDS-PAGE), buffered aqueous solution, ≥30,000 units/mg protein
C9284 Catalase−Agarose, pH 5.4(isoelectric point), ammonium sulfate suspension, enzyme from bovine liver
L2005 Lactoperoxidase from bovine milk, lyophilized powder (essentially salt-free), ≥200 units/mg protein
61328 Lactoperoxidase from bovine milk, lyophilized, powder, ≥150 U/mg
M6908 Myeloperoxidase from human leukocytes, lyophilized powder, ≥50 units/mg protein
68528 Peroxidase from Bjerkandera adusta, ≥2.8 U/mg
P8375 Peroxidase from horseradish, Type VI, essentially salt-free, lyophilized powder, ≥250 units/mg solid (using pyrogallol)
77332 Peroxidase from horseradish, lyophilized, powder, beige, ~150 U/mg
P6782 Peroxidase from horseradish, Type VI-A, essentially salt-free, lyophilized powder, 950-2000 units/mg solid (using ABTS), ≥250 units/mg solid (using pyrogallol)
P8250 Peroxidase from horseradish, Type II, essentially salt-free, lyophilized powder, 150-250 units/mg solid (using pyrogallol)
P8125 Peroxidase from horseradish, Type I, essentially salt-free, lyophilized powder, 50-150 units/mg solid (using pyrogallol)
P8415 Peroxidase from horseradish, Type XII, essentially salt-free, lyophilized powder, ≥250 units/mg solid (using pyrogallol)
P6140 Peroxidase from horseradish, Type X, ammonium sulfate suspension
P2088 Peroxidase from horseradish, Highly stabilized, essentially salt-free, lyophilized powder, 200-300 units/mg solid (using pyrogallol)
T3824
Tyrosinase from mushroom, lyophilized powder, ≥1000 unit/mg solid