EMAIL THIS PAGE TO A FRIEND
The FEBS journal
Multimeric and differential binding of CIN85/CD2AP with two atypical proline-rich sequences from CD2 and Cbl-b*.
PMID 23663663
Abstract
The CD2AP (CD2-associated protein) and CIN85 (Cbl-interacting protein of 85 kDa) adaptor proteins each employ three Src homology 3 (SH3) domains to cluster protein partners and ensure efficient signal transduction and down-regulation of tyrosine kinase receptors. Using NMR, isothermal titration calorimetry and small-angle X-ray scattering methods, we have characterized several binding modes of the N-terminal SH3 domain (SH3A) of CD2AP and CIN85 with two natural atypical proline-rich regions in CD2 (cluster of differentiation 2) and Cbl-b (Casitas B-lineage lymphoma), and compared these data with previous studies and published crystal structures. Our experiments show that the CD2AP-SH3A domain forms a type II dimer with CD2 and both type I and type II dimeric complexes with Cbl-b. Like CD2AP, the CIN85-SH3A domain forms a type II complex with CD2, but a trimeric complex with Cbl-b, whereby the type I and II interactions take place at the same time. Together, these results explain how multiple interactions among similar SH3 domains and ligands produce a high degree of diversity in tyrosine kinase, cell adhesion or T-cell signaling pathways.