EMAIL THIS PAGE TO A FRIEND
Journal of bioscience and bioengineering
Biochemical characterization and cooperation with co-chaperones of heat shock protein 90 from Schizosaccharomyces pombe.
PMID 23664927
Abstract
The characterization of Hsp90 from the fission yeast Schizosaccharomyces pombe was performed. Hsp90 of S. pombe existed as a dimer and exhibited ATP-dependent conformational changes. It captured unfolded proteins in the ATP-free open conformation and protected them from thermal aggregation. Hsp90 of S. pombe was also able to refold thermally denatured firefly luciferase. The co-chaperones Sti1 and Aha1 bound Hsp90 and modulated its activity. Because the affinity of Sti1 was higher than that of Aha1, the effect of Sti1 appeared to dominate when both co-chaperones existed simultaneously.