EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica
[Chemical constituents of Rhododendron seniavinii].
PMID 23668011
Abstract
To study the chemical constituents of Rhododendron seniavinii. Compounds were isolated from the aqueous extract of the leaves of R. seniavinii by using Sephadex LH-20, ODS open column chromatography and other means. Their structures were elucidated according to spectral data and physiochemical properties. Thirteen compounds were isolated from R. seniavinii and identified as 5-methoxydehydroconiferyl alcohol (1), dehydroconiferyl alcohol (2), (-)-syringaresinol (3), (-)-lyoniresinol (4), (+)-lyoniresinol 3alpha-O-beta-D-glucopyranoside (5), (-)-lyoniresinol 3alpha-O-beta-D-glucopyranoside (6), 3,4,5-trimethoxyphenyl-1-O-beta-D-glucopyranoside (7), nikoenoside (8), 3,5,7-trihydroxychromone-3-0-alpha-L-rhamnopyranoside (9), 3,4,5-trimethoxyphenol (10), scopoletin (11), scopolin (12) and quercitrin (13). Compounds 1-12 were obtained from this plant for the first time.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
00268 Dextran, analytical standard, for GPC, 1,000
00269 Dextran, analytical standard, for GPC, 5,000
00270 Dextran, analytical standard, for GPC, 12,000
00271 Dextran, analytical standard, for GPC, 25,000
00891 Dextran, analytical standard, for GPC, 50,000
00892 Dextran, analytical standard, for GPC, 80,000
00893 Dextran, analytical standard, for GPC, 150,000
00894 Dextran, analytical standard, for GPC, 270,000
00895 Dextran, analytical standard, for GPC, 410,000
00896 Dextran, analytical standard, for GPC, 670,000
31394 Dextran, enzymatic synth.
31430 Dextran, analytical standard, for GPC, Set Mp 1,000-400,000
31416 Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 1,000
31417 Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 5,000
D9260 Dextran from Leuconostoc mesenteroides, average mol wt 9,000-11,000
31418 Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 12,000
D4626 Dextran from Leuconostoc mesenteroides, average mol wt 15,000-30,000
31419 Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 25,000
D1662 Dextran from Leuconostoc mesenteroides, average mol wt 35,000-45,000
31420 Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 50,000
D3759 Dextran from Leuconostoc mesenteroides, average mol wt 48,000-90,000
31421 Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 80,000
D4876 Dextran from Leuconostoc mesenteroides, average mol wt 150,000
31422 Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 150,000
31423 Dextran from Leuconostoc mesenteroides, for GPC, Mw 270,000
31424 Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 410,000
31425 Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 670,000
D5376 Dextran from Leuconostoc mesenteroides, average mol wt 1,500,000-2,800,000
31397 Dextran from Leuconostoc mesenteroides, Mr ~60,000
31398 Dextran from Leuconostoc mesenteroides, Mr ~200,000