EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica
[Protective effect of medicated serum prepared with taohong siwu tang on hydrogen peroxide-induced human umbilical vein endothelial cells].
PMID 23668018
Abstract
To study the protective effect of medicated serum prepared with Taohong Siwu Tang on hydrogen peroxide-injured human umbilical vein endothelial cells (HUVECs). Sprague Dawley rats were orally administered with 10 mL x kg(-1) extracts from Taohong Siwu Tang (1.75 g crude drug), twice a day for three days, in order to prepare medicated serum of Taohong Siwu Tang. The effect of medicated serum pre-treated with Taohong Siwu Tang (with concentrations of 5%, 10%, 15%) on reduction of H2O2-induced cell activity was detected MTT. Cell morphological changes were observed under microscope. The effect of medicated serum prepared with Taohong Siwu Tang on the apoptosis and antioxidant capacity of HUVECs was detected with the content of malondialdehyde (MDA) and dehydrogenase (LDH), the activity of superoxide dismutase (SOD) and AO/EB staining. The expression of Caspase-3 was determined by western blot. Compared with the blank serum control group, cell were significantly less active after being damaged by H2O2 (400 micromol x L(-1)). Medicated serum could significantly improve the SOD activity, reduce the levels of LDH and MDA, and inhibite the expression of Caspase-3. AO/EB staining and inverted microscope also showed that medicated serum prepared with Taohong Siwu Tang could reduce cell apoptosis induced by H2O2. Medicated serum prepared with Taohong Siwu Tang has significant protective effect on HUVECs injured by H2O2.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L3916 L-Lactic Dehydrogenase from bovine heart, 1000 units/mL
L2625 L-Lactic Dehydrogenase from bovine heart, Type III, ammonium sulfate suspension, ≥500 units/mg protein
L1006 L-Lactic Dehydrogenase from bovine heart, Type XVII, buffered aqueous glycerol solution, ≥400 units/mg protein
L1378 L-Lactic Dehydrogenase from bovine muscle, Type X, ammonium sulfate suspension, ≥600 units/mg protein
L7525 L-Lactic Dehydrogenase from porcine heart, ammonium sulfate suspension, ≥200 units/mg protein
L2500 L-Lactic Dehydrogenase from rabbit muscle, Type II, ammonium sulfate suspension, 800-1,200 units/mg protein
L1254 L-Lactic Dehydrogenase from rabbit muscle, Type XI, lyophilized powder, 600-1,200 units/mg protein
59747 Lactic Dehydrogenase, recombinant from E. coli, ≥90 U/mg
S9697
Superoxide Dismutase bovine, recombinant, expressed in E. coli, lyophilized powder, ≥2500 units/mg protein, ≥90% (SDS-PAGE)
S5395
Superoxide Dismutase from bovine erythrocytes, BioReagent, ≥3,000 units/mg protein, suitable for cell culture, lyophilized powder
S7571
Superoxide Dismutase from bovine erythrocytes, lyophilized powder, ≥3,000 units/mg protein
S7446
Superoxide Dismutase from bovine erythrocytes, BioUltra, lyophilized powder, ≥4,500 units/mg protein, ≥97% (SDS-PAGE)
S8160
Superoxide Dismutase from bovine liver, lyophilized powder, ≥1500 units/mg protein
S8409
Superoxide Dismutase from bovine liver, ammonium sulfate suspension, 2,000-6,000 units/mg protein (biuret)
S5639
Superoxide Dismutase from Escherichia coli, lyophilized powder, ≥1,000 units/mg protein
S9636
Superoxide Dismutase from human erythrocytes, essentially salt-free, lyophilized powder, ≥2,500 units/mg protein
S9076 Superoxide Dismutase I human, recombinant, expressed in E. coli