EMAIL THIS PAGE TO A FRIEND
Profiles of drug substances, excipients, and related methodology
Paroxetine hydrochloride.
PMID 23668408
Abstract
Paroxetine hydrochloride (3S-trans)-3-[(1,3-benzodioxol-5-yloxy)methyl]-4-(4-fluorophenyl)-piperidine hydrochloride (or (-)-(3S,4R)-(4-(p-fluorophenyl)-3-[[3,4-(methylenedioxy)-phenoxy]methyl]piperidine hydrochloride), a phenylpiperidine derivative, is a selective serotonin reuptake inhibitor. Paroxetine is indicated for the treatment of depression, generalized anxiety disorder, obsessive-compulsive disorder, panic disorder, posttraumatic stress disorder, and social anxiety disorder. The physicochemical properties, spectroscopic data (1D and 2D NMR, UV, FT-IR, MS, PXRD), stability, methods of preparation and chromatographic methods of analysis of pharmaceutical, and biological samples of paroxetine are documented in this review. Pharmacokinetics, metabolism, and pharmacological effects are also discussed.