EMAIL THIS PAGE TO A FRIEND
Pediatrics
Lyme chondritis presenting as painless ear erythema.
PMID 23669522
Abstract
Erythema of the ear lobe in the context of Lyme disease is caused by either borrelial lymphocytoma or localized erythema migrans. Here we present a case of chondritis limited to the ear cartilage caused by Lyme disease. The patient was treated with ceftriaxone with complete resolution of symptoms.
Related Materials