EMAIL THIS PAGE TO A FRIEND
Journal of pediatric hematology/oncology
Successful treatment of recurrent pediatric inflammatory myofibroblastic tumor in a single patient with a novel chemotherapeutic regimen containing celecoxib.
PMID 23669730
Abstract
Inflammatory myofibroblastic tumors are rare tumors characterized as low-to-intermediate grade sarcomas. This is a case of a 7-year-old male with a 5-cm lung mass, which recurred 11 months after complete resection. The recurrence manifested as multifocal metastatic disease involving the ipsilateral parietal and visceral pleura. A novel chemotherapeutic regimen, which included vincristine, ifosfamide, doxorubicin, and celecoxib was utilized for the disease recurrence. The patient had complete and durable remission of the disease and has been disease-free for >4 years. This novel regimen including a cyclooxygenase 2 inhibitor may be an effective regimen for metastatic inflammatory myofibroblastic tumors.