EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
182Hf-182W age dating of a 26Al-poor inclusion and implications for the origin of short-lived radioisotopes in the early Solar System.
PMID 23671077
Abstract
Refractory inclusions [calcium-aluminum-rich inclusions, (CAIs)] represent the oldest Solar System solids and provide information regarding the formation of the Sun and its protoplanetary disk. CAIs contain evidence of now extinct short-lived radioisotopes (e.g., (26)Al, (41)Ca, and (182)Hf) synthesized in one or multiple stars and added to the protosolar molecular cloud before or during its collapse. Understanding how and when short-lived radioisotopes were added to the Solar System is necessary to assess their validity as chronometers and constrain the birthplace of the Sun. Whereas most CAIs formed with the canonical abundance of (26)Al corresponding to (26)Al/(27)Al of ∼5 × 10(-5), rare CAIs with fractionation and unidentified nuclear isotope effects (FUN CAIs) record nucleosynthetic isotopic heterogeneity and (26)Al/(27)Al of <5 × 10(-6), possibly reflecting their formation before canonical CAIs. Thus, FUN CAIs may provide a unique window into the earliest Solar System, including the origin of short-lived radioisotopes. However, their chronology is unknown. Using the (182)Hf-(182)W chronometer, we show that a FUN CAI recording a condensation origin from a solar gas formed coevally with canonical CAIs, but with (26)Al/(27)Al of ∼3 × 10(-6). The decoupling between (182)Hf and (26)Al requires distinct stellar origins: steady-state galactic stellar nucleosynthesis for (182)Hf and late-stage contamination of the protosolar molecular cloud by a massive star(s) for (26)Al. Admixing of stellar-derived (26)Al to the protoplanetary disk occurred during the epoch of CAI formation and, therefore, the (26)Al-(26)Mg systematics of CAIs cannot be used to define their formation interval. In contrast, our results support (182)Hf homogeneity and chronological significance of the (182)Hf-(182)W clock.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
11009
Aluminum, powder, ≥91% (complexometric)
Al
518573
Aluminum, granular, <1 mm, 99.7% trace metals basis
Al
266574
Aluminum, foil, thickness 0.45-0.55 mm, 99.999% trace metals basis
Al
653608
Aluminum, powder, <5 μm particle size, 99.5% trace metals basis
Al
266558
Aluminum, wire, diam. 1.0 mm, 99.999% trace metals basis
Al
266523
Aluminum, ReagentPlus®, beads, 5-15 mm, 99.9% trace metals basis
Al
202584
Aluminum, powder, <75 μm, ≥99.95% trace metals basis
Al
338788
Aluminum, pellets, 3-12 mm, 99.99% trace metals basis
Al
433705
Aluminum, evaporation slug, diam. × L 6.3 mm × 6.3 mm, 99.999% trace metals basis
Al
310360
Aluminum, ACS reagent, 99%, wire, wire diam. ~1.5 mm
Al
326852
Aluminum, foil, thickness 0.25 mm, 99.999% trace metals basis
Al
266957
Aluminum, foil, thickness 1.0 mm, 99.999% trace metals basis
Al
266515
Aluminum, ≤30 μm, spherical powder, ≥99%
Al
326860
Aluminum, foil, thickness 0.13 mm, ≥99.99% trace metals basis
Al
733369
Aluminum, foil, thickness 8 μm, 99% trace metals basis
Al
326887
Aluminum, wire, diam. 0.58 mm, ≥99.99% trace metals basis
Al
326941
Aluminum, pellets, 3-8 mesh, ≥99.999% trace metals basis
Al
749036
Aluminum, sputtering target, diam. × thickness 3.00 in. × 0.125 in., 99.9995% trace metals basis
Al
202576
Aluminum, rod, diam. × L 3.0 mm × 100 mm, 99.999% trace metals basis
Al
IRMM523B
Aluminum, IRMM® certified Reference Material, 1.0 mm foil
Al
IRMM523A
Aluminum, IRMM® certified Reference Material, 0.1 mm foil
Al
IRMM523C
Aluminum, IRMM® certified Reference Material, 1.0 mm wire
Al
266752
Hafnium, powder, −325 mesh, 99.5% trace metals basis (purity excludes ~2% zirconium), contains 1:10 pentanol to water solution as stabilizer
Hf
266809
Hafnium, turnings, crystal bar, 99.7% trace metals basis
Hf
267511
Tungsten, powder, 12 μm, 99.9% trace metals basis
W
267538
Tungsten, foil, thickness 0.05 mm, ≥99.9% trace metals basis
W
357421
Tungsten, powder, 10 μm, ≥99.99% trace metals basis
W
267554
Tungsten, wire, diam. 0.25 mm, ≥99.9% trace metals basis
W
510106
Tungsten, powder (monocrystalline), 0.6-1 μm, ≥99.9% trace metals basis
W
267546
Tungsten, foil, thickness 0.25 mm, ≥99.9% trace metals basis
W