EMAIL THIS PAGE TO A FRIEND
Journal of the College of Physicians and Surgeons--Pakistan : JCPSP
Focal dystonia of right hand with mirror movements upon use of left arm.
PMID 23673180
Abstract
Dystonia is a movement disorder characterized by sustained muscle contractions, causing twisting and repetitive movements or abnormal postures of affected body parts. Here, we present a novel case of focal dystonia of a 51 years old right-handed woman who had developed difficulty in writing and performing fine motor tasks. Due to a discomfort in her right hand at use, she started using her left hand instead and noticed inconsistent mirror movements in her right hand upon use of left hand. She was treated with trihexyphenidyl which allowed her right hand to function better, though writing still remained a problem.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000108
Trihexyphenidyl hydrochloride, European Pharmacopoeia (EP) Reference Standard
C20H31NO · HCl