EMAIL THIS PAGE TO A FRIEND
European journal of drug metabolism and pharmacokinetics
Is pomegranate juice a potential perpetrator of clinical drug-drug interactions? Review of the in vitro, preclinical and clinical evidence.
PMID 23673492
Abstract
The area of fruit juice-drug interaction has received wide attention with numerous scientific and clinical investigations performed and reported for scores of drugs metabolized by CYP3A4/CYP2C9. While grapefruit juice has been extensively studied with respect to its drug-drug interaction potential, numerous other fruit juices such as cranberry juice, orange juice, grape juice, pineapple juice and pomegranate juice have also been investigated for its potential to show drug-drug interaction of any clinical relevance. This review focuses on establishing any relevance for clinical drug-drug interaction potential with pomegranate juice, which has been shown to produce therapeutic benefits over a wide range of disease areas. The review collates and evaluates relevant published in vitro, preclinical and clinical evidence of the potential of pomegranate juice to be a perpetrator in drug-drug interactions mediated by CYP3A4 and CYP2C9. In vitro and animal pharmacokinetic data support the possibility of CYP3A4/CYP2C9 inhibition by pomegranate juice; however, the human relevance for drug-drug interaction was not established based on the limited case studies.