EMAIL THIS PAGE TO A FRIEND
Journal of computer assisted tomography
Comparison of multidetector computed tomography angiography and cholangiography performed at 80 and 120 kVp in live liver donors.
PMID 23674019
Abstract
The objective of this study was to compare the radiation exposure and image quality of contrast-enhanced multidetector computed tomography angiography (CTA) and computed tomography cholangiography (CTC) performed for living liver donor evaluation using 80 and 120 kVp. Ninety-three potential liver donors who underwent preoperative contrast-enhanced 64 multidetector CTA and CTC were retrospectively divided into 2 groups: at 80 and at 120 kVp. An institutional review board waiver was obtained. Signal-to-noise ratio and contrast-to-noise ratio of the hepatic artery and common bile duct were obtained. The dose-length product was recorded. Image quality and visibility of hepatic artery and biliary tract anatomy were evaluated. Mann-Whitney U test was used for statistical evaluation. Mean hepatic artery/common bile duct signal-to-noise ratio was 28.9/28.6 (SD, 14.2/10.0) at 80 kVp and 27.6/25.8 (SD, 8.0/6.2) at 120 kVp (P = 0.61/0.099). Mean hepatic artery/common bile duct contrast-to-noise ratio was 24.8/23.3 (SD, 12.9/8.6) at 80 kVp and 22.2/19.3 (SD, 7.7/5.0) at 120 kVp (P = 0.76/0.005). Mean CTA/CTC dose-length product was 279/281 (SD, 42/52) mGy-cm at 80 kVp and 407/451 (SD, 208/243) mGy-cm at 120 kVp (P = 0.026/0.002). Computed tomography cholangiography image quality and visibility of biliary tract anatomy were not significantly different at 80 versus 120 kVp (all P > 0.13). Computed tomography angiography image quality was significantly lower (P < 0.01), and the noise scores significantly higher (P < 0.01) at 80 versus 120 kVp, but diagnostic. Contrast-enhanced CTA and CTC performed at 80 kVp result in comparable image quality and anatomical evaluation with reduced radiation exposure when compared with 120 kVp.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I0639
Iodipamide, analytical standard, for drug analysis
C20H14I6N2O6