EMAIL THIS PAGE TO A FRIEND
The Analyst
Use of a large Stokes-shift fluorophore to increase the multiplexing capacity of a point-of-care DNA diagnostic device.
PMID 23675581
Abstract
The intense demand for fluorescence-based point of care (POC) DNA diagnostics is driving developments to reduce the size of instrumentation, imposing limitations on the optical hardware that can be included. Here we describe a combination of instrumentation and fluorogenic probes to detect three fluorophores using two excitation and two detection channels.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
93543
N-Hydroxysuccinimide (NHS) activated MegaStokes dye 605
C24H28F6N3O4P