EMAIL THIS PAGE TO A FRIEND
Journal of helminthology
Tandem antioxidant enzymes confer synergistic protective responses in experimental filariasis.
PMID 23676147
Abstract
Helminth parasites use antioxidant defence strategies for survival during oxidative stress due to free radicals in the host. Accordingly, tissue-dwelling filarial parasites counteract host responses by releasing a number of antioxidants. Targeting these redox regulation proteins together, would facilitate effective parasite clearance. Here, we report the combined effect of protective immune responses trigged by recombinant Wuchereria bancrofti thioredoxin (WbTRX) and thioredoxin peroxidase (WbTPX) in an experimental filarial model. The expression of WbTRX and WbTPX in different stages of the parasite and their cross-reactivity were analysed by enzyme-linked immunosorbent assay (ELISA). The immunogenicity of recombinant proteins and their protective efficacy were studied in animal models when immunized in single or cocktail mode. The antigens showed cross-reactive epitopes and induced high humoral and cellular immune responses in mice. Further, parasite challenge against Brugia malayi L3 larvae in Mastomys coucha conferred significant protection of 57% and 62% against WbTRX and WbTPX respectively. The efficacy of L3 clearance was significantly higher (71%) (P <  0.001) when the antigens were immunized together, showing a synergistic effect in multiple-mode vaccination. Hence, the study suggests WbTRX and WbTPX to be attractive vaccine candidates when immunized together and provides a tandem block for parasite elimination in the control of lymphatic filariasis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T0803
Anti-Thioredoxin antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB2501722
Anti-TPO antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1406549
Anti-TPO antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA007987
Anti-TPO antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1408638
Anti-TPO antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1409783
Anti-TXN antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB4200074
Anti-VAC14 antibody produced in rabbit, ~1.5 mg/mL, affinity isolated antibody, buffered aqueous solution
HPA027766
Anti-VAC14 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB5300166
Monoclonal Anti-Thioredoxin (TRX) antibody produced in mouse, clone 1H6H6, ascites fluid
SAB1402382
Monoclonal Anti-TPO antibody produced in mouse, clone 2A11, purified immunoglobulin, buffered aqueous solution
SAB1404501
Monoclonal Anti-TXN antibody produced in mouse, clone 6C10, purified immunoglobulin, buffered aqueous solution
WH0007295M1
Monoclonal Anti-TXN antibody produced in mouse, clone 2A7, purified immunoglobulin, buffered aqueous solution
SAB1401760
Monoclonal Anti-VAC14 antibody produced in mouse, clone 3B2, purified immunoglobulin, buffered aqueous solution
SAB1400627
Anti-VAC14 antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
SAB2103378
Anti-SASP antibody produced in rabbit, affinity isolated antibody
SAB1411371
Anti-TXN antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution