EMAIL THIS PAGE TO A FRIEND
The Journal of neuroscience : the official journal of the Society for Neuroscience
Ligand for translocator protein reverses pathology in a mouse model of Alzheimer's disease.
PMID 23678130
Abstract
Ligands of the translocator protein (TSPO) elicit pleiotropic neuroprotective effects that represent emerging treatment strategies for several neurodegenerative conditions. To investigate the potential of TSPO as a therapeutic target for Alzheimer's disease (AD), the current study assessed the effects of the TSPO ligand Ro5-4864 on the development of neuropathology in 3xTgAD mice. The effects of the TSPO ligand on neurosteroidogenesis and AD-related neuropathology, including β-amyloid accumulation, gliosis, and behavioral impairment, were examined under both early intervention (7-month-old young-adult male mice with low pathology) and treatment (24-month-old, aged male mice with advanced neuropathology) conditions. Ro5-4864 treatment not only effectively attenuated development of neuropathology and behavioral impairment in young-adult mice but also reversed these indices in aged 3xTgAD mice. Reduced levels of soluble β-amyloid were also observed by the combination of TSPO ligands Ro5-4864 and PK11195 in nontransgenic mice. These findings suggest that TSPO is a promising target for the development of pleiotropic treatment strategies for the management of AD.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C5174
4′-Chlorodiazepam, ≥98% (TLC)
C16H12Cl2N2O