EMAIL THIS PAGE TO A FRIEND
Natural product communications
Chemical constituents from Dendropanax dentiger.
PMID 23678812
Abstract
Nineteen compounds and an HPLC inseparable mixture, composed of compounds 20 and 21, were isolated from the leaves and twigs of Dendropanax dentiger (Harms ex Diels) Merr. Of these, syringin (1) is the most abundant, 6'-O-apiofuranosyl dendranthemoside A (16) is a new megastigmane glycoside, and 3-methoxy-D-mannono-1,4-lactone (21) is a new hexono-1,4-lactone. Their structures were elucidated based on NMR spectroscopic and MS analyses.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
90974
Eleutheroside B, analytical standard
C17H24O9