EMAIL THIS PAGE TO A FRIEND
Clinical science (London, England : 1979)
Human parainfluenza type 3 virus impairs the efficacy of glucocorticoids to limit allergy-induced pulmonary inflammation in guinea-pigs.
PMID 23678868
Abstract
Viral exacerbations of allergen-induced pulmonary inflammation in pre-clinical models reportedly reduce the efficacy of glucocorticoids to limit pulmonary inflammation and airways hyper-responsiveness to inhaled spasmogens. However, exacerbations of airway obstruction induced by allergen challenge have not yet been studied. hPIV-3 (human parainfluenza type 3 virus) inoculation of guinea-pigs increased inflammatory cell counts in BAL (bronchoalveolar lavage) fluid and caused hyper-responsiveness to inhaled histamine. Both responses were abolished by treatment with either dexamethasone (20 mg/kg of body weight, subcutaneous, once a day) or fluticasone propionate (a 0.5 mg/ml solution aerosolized and inhaled over 15 min, twice a day). In ovalbumin-sensitized guinea-pigs, allergen (ovalbumin) challenge caused two phases of airway obstruction [measured as changes in sGaw (specific airways conductance) using whole body plethysmography]: an immediate phase lasting between 4 and 6 h and a late phase at about 7 h. The late phase, airway hyper-responsiveness to histamine and inflammatory cell counts in BAL were all significantly reduced by either glucocorticoid. Inoculation of guinea-pigs sensitized to ovalbumin with hPIV-3 transformed the allergen-induced airway obstruction from two transient phases into a single sustained response lasting up to 12 h. This exacerbated airway obstruction and airway hyper-responsiveness to histamine were unaffected by treatment with either glucocorticoid whereas inflammatory cell counts in BAL were only partially inhibited. Virus- or allergen-induced pulmonary inflammation, individually, are glucocorticoid-sensitive, but in combination generate a phenotype where glucocorticoid efficacy is impaired. This suggests that during respiratory virus infection, glucocorticoids might be less effective in limiting pulmonary inflammation associated with asthma.