EMAIL THIS PAGE TO A FRIEND
The Journal of thoracic and cardiovascular surgery
Use of annulus washer after debridement: a new mitral valve replacement technique for patients with severe mitral annular calcification.
PMID 23679966