EMAIL THIS PAGE TO A FRIEND
Environmental science. Processes & impacts
Passive monitors to measure hydrogen sulfide near concentrated animal feeding operations.
PMID 23681048
Abstract
Hydrogen sulfide (H2S) is one of many airborne pollutants emitted by concentrated animal feeding operations (CAFOs). However, few studies have characterized ambient H2S levels near these facilities, largely due to the lack of low-cost, reliable, and easily transportable instrumentation available to researchers. We determined intermediate environmental H2S exposure near CAFOs using Radiello passive monitors. First, a laboratory study was performed to determine the accuracy of the device. Next, a total of eight passive H2S monitors were deployed bi-weekly in close proximity (<40 m) to a medium-sized swine confinement for seven months in order to determine the temporal and spatial variability of H2S. Finally, we measured H2S concentrations across two rural Iowa counties to characterize ambient exposure near thirteen CAFOs and two schools. The value of the temperature-adjusted H2S passive diffusion rate provided by the supplier was 29% larger than the 24 h rate determined experimentally. Concentrations of H2S measured near the medium-sized confinement were varied and ranged from 0.2 to 48.6 ppb depending on the sampling period and proximity to a lagoon on the property. Two-week concentrations near the schools were low (<1 ppb), while concentrations near the thirteen CAFOs ranged from 0.1 to 42.9 ppb. The passive monitors were effective in measuring H2S concentrations near a swine CAFO as long as they were exposed for a sufficient period of time (two weeks). Radiello passive monitors are a promising new device in measuring intermediate H2S exposure in rural populations. Measured values in excess of an Iowa state limit of 30 ppb (24 h average) suggest that enforcement actions are needed to mitigate H2S migration from swine CAFOs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
RAD141S
radiello 1,3-Butadiene Starter Kit, Thermal Desorption, pkg of 1 kit (2 RAD141 adsorbing cartridges and barcode labels, 1 RAD1202 yellow diffusive body, 1 RAD121 triangular support plate, 1 instruction sheet)
RAD125
radiello Anesthetic Gases and Vapor Sampler
RAD190
radiello Barcode Labels, pk of 198
RAD405
radiello BTEX Calibration Kit (CS2 Desorption)
RAD407 radiello BTEX Calibration Kit (Thermal Desorption)
RAD130S
radiello BTEX/VOC Starter Kit, CS2 Desorption
RAD145S
radiello BTEX/VOC Starter Kit, Thermal Desorption
RAD165
radiello Cartridge Adsorbents, for sampling Aldehydes, matrix SS net with 2,4-DNPH coated FLORISIL®, pk of 20
RAD168
radiello Cartridge Adsorbents, for sampling Ammonia (NH3), matrix microporous PE impregnated with phosphoric acid, pk of 20
RAD132
radiello Cartridge Adsorbents, for sampling Anaesthetic Gases/Vapors, matrix SS net with mix of mol sieve and activated charcoal (30-50 mesh), pk of 20
RAD130
radiello Cartridge Adsorbents, for sampling BTEX and VOCs (CS2 Desorption), matrix SS net (100 mesh, 5.8 mm diam.), activated with activated charcoal (30-50 mesh), pk of 20
RAD145
radiello Cartridge Adsorbents, for sampling BTEX and VOCs (thermal desorption), matrix SS net (3 x 8 μm, 4.8 mm diam.), Carbograph, pk of 20
RAD195 radiello Clips, suspending Radiello triangular support plate, pk of 20
RAD120
radiello Diffusive Bodies, white, configured for general use, pk of 20
RAD1201
radiello Diffusive Bodies, blue, configured for sampling light sensitive compounds, pk of 20
RAD1202
radiello Diffusive Bodies, yellow, configured for reduced sampling rates, pk of 20
RAD1203
radiello Diffusive Bodies, permeative, configured for anaesthetic gases and vapors, pk of 20
RAD1991
radiello Empty Storage Tubes, glass (with stopper), volume 2.8 mL, pk of 100
RAD1992
radiello Empty Storage Tubes, polypropylene (with stopper), volume 12 mL, pk of 100
RAD174
radiello Filtration Kit, pk of 20
RAD170S
radiello Hydrogen Sulfide Starter Kit
RAD171 radiello Methylene Blue Calibration Standard for H2S (Hydrogen Sulfide), pkg of 100 mL
RAD126
radiello On-Field Thermometer, Includes one standard Radiello vertical adapter (RAD122) for each thermometer, configured for standard use, pk of 3
RAD1261
radiello On-Field Thermometer, configured for ready-to-use (rtu) samplers, Include one rtu vertical adapter for each thermometer, pk of 3
RAD127
radiello On-Field Thermometer, Reader, pk of 1
RAD196
radiello Outdoor Shelter, pk of 10
RAD198 radiello Replacement Mounting Strips, for use with Radiello Outdoor Shelter, pk of 100
RAD121
radiello Triangular Support Plate, pk of 20
RAD122
radiello Vertical Adapter, threaded for standard use, pk of 20
RAD1221
radiello Vertical Adapter, non-threaded for ready-to-use samplers, pk of 20