EMAIL THIS PAGE TO A FRIEND
Undersea & hyperbaric medicine : journal of the Undersea and Hyperbaric Medical Society, Inc
Oxidative stress, antioxidant defenses and nitric oxide production following hyperoxic exposures.
PMID 23682544
Abstract
Little data exist on the dose-response relationship between the partial pressure of inspired oxygen (PiO2) and the cellular oxidative stress response in humans. The objective of this study was to determine the effects of PiO2 on lipid peroxidation, antioxidant enzyme activity and nitric oxide (NO) production. Twelve healthy male divers breathed 100% O2 in a hyperbaric chamber for two hours at 1 (101 kPa), 1.5 (152 kPa) and 2 (203 kPa) atmospheres absolute (atm abs). Venous blood was collected pre-, within 15 minutes post-, one and two hours post-hyperoxic exposures to determine changes in plasma and erythrocyte lipid peroxidation (thiobarbituric acid reactive substances-TBARS), antioxidant enzyme activity (superoxide dismutase-SOD), catalase-CAT, glutathione peroxidase-GPx), and plasma NO production (L-arginine [L-Arg], asymmetric dimethylarginine-ADMA, and nitrites). There were minor changes in TBARS and mixed responses in plasma and erythrocyte CAT and GPx activity. Plasma L-Arg increased following 1 and 1.5 atm abs exposures, yet ADMA and nitrites were unchanged. Only erythrocyte CAT and plasma GPx activity, and plasma L-Arg/ADMA demonstrated a significant PiO2 dose-dependent relationship. Two-hour hyperoxic exposures at 1-2 atm abs of O2 results in mixed oxidant-antioxidant responses and unaltered NO production. Moreover, there does not appear to be a strong systemic dose-dependent oxidative stress response at these hyperoxic exposures.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C3515 Catalase from Aspergillus niger, ammonium sulfate suspension, ≥4,000 units/mg protein
02071 Catalase from Corynebacterium glutamicum, solution, deep brown, ≥500000 U/mL
C3556 Catalase from human erythrocytes, ≥90% (SDS-PAGE), buffered aqueous solution, ≥30,000 units/mg protein
C9284 Catalase−Agarose, pH 5.4(isoelectric point), ammonium sulfate suspension, enzyme from bovine liver
G6137 Glutathione Peroxidase from bovine erythrocytes, lyophilized powder, ≥300 units/mg protein
S9697
Superoxide Dismutase bovine, recombinant, expressed in E. coli, lyophilized powder, ≥2500 units/mg protein, ≥90% (SDS-PAGE)
S5395
Superoxide Dismutase from bovine erythrocytes, ≥3,000 units/mg protein, BioReagent, lyophilized powder, suitable for cell culture
S7446
Superoxide Dismutase from bovine erythrocytes, BioUltra, lyophilized powder, ≥4,500 units/mg protein, ≥97% (SDS-PAGE)
S8160
Superoxide Dismutase from bovine liver, lyophilized powder, ≥1500 units/mg protein
S8409
Superoxide Dismutase from bovine liver, ammonium sulfate suspension, 2,000-6,000 units/mg protein (biuret)
S5639
Superoxide Dismutase from Escherichia coli, lyophilized powder, ≥1,000 units/mg protein
S9636
Superoxide Dismutase from human erythrocytes, essentially salt-free, lyophilized powder, ≥2,500 units/mg protein
S9076 Superoxide Dismutase I human, recombinant, expressed in E. coli