EMAIL THIS PAGE TO A FRIEND
Hepatology (Baltimore, Md.)
The receptor TGR5 protects the liver from bile acid overload during liver regeneration in mice.
PMID 23686672
Abstract
Many regulatory pathways are involved in liver regeneration after partial hepatectomy (PH) to initiate growth, protect liver cells, and sustain functions of the remnant liver. Bile acids (BAs), whose levels rise in the blood early after PH, stimulate both hepatocyte proliferation and protection, in part through their binding to the nuclear farnesoid X receptor (FXR). However, the effect of the BA receptor, TGR5 (G-protein-coupled BA receptor 1) after PH remains to be studied. Liver histology, hepatocyte proliferation, BA concentrations (plasma, bile, liver, urine, and feces), bile flow and composition, and cytokine production were studied in wild-type (WT) and TGR5 KO (knockout) mice before and after PH. BA composition (plasma, bile, liver, urine, and feces) was more hydrophobic in TGR5 KO than in WT mice. After PH, severe hepatocyte necrosis, prolonged cholestasis, exacerbated inflammatory response, and delayed regeneration were observed in TGR5 KO mice. Although hepatocyte adaptive response to post-PH BA overload was similar in WT and TGR5 KO mice, kidney and biliary adaptive responses were strongly impaired in TGR5 KO mice. Cholestyramine treatment, as well as Kupffer cell depletion, significantly improved the post-PH TGR5 KO mice phenotype. After bile duct ligation or upon a cholic acid-enriched diet, TGR5 KO mice exhibited more severe liver injury than WT as well as impaired BA elimination in urine. TGR5 is crucial for liver protection against BA overload after PH, primarily through the control of bile hydrophobicity and cytokine secretion. In the absence of TGR5, intrahepatic stasis of abnormally hydrophobic bile and excessive inflammation, in association with impaired bile flow adaptation and deficient urinary BA efflux, lead to BA overload-induced liver injury and delayed regeneration.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C4650 Cholestyramine resin