EMAIL THIS PAGE TO A FRIEND
Journal of Molecular and Cellular Cardiology
ADAM17 silencing by adenovirus encoding miRNA-embedded siRNA revealed essential signal transduction by angiotensin II in vascular smooth muscle cells.
PMID 23688779