EMAIL THIS PAGE TO A FRIEND
Otolaryngology--head and neck surgery : official journal of American Academy of Otolaryngology-Head and Neck Surgery
Sensorineural hearing loss associated with neomycin eardrops and nonintact tympanic membranes.
PMID 23690082