EMAIL THIS PAGE TO A FRIEND
Otolaryngology--head and neck surgery : official journal of American Academy of Otolaryngology-Head and Neck Surgery
Possible errors in hearing recovery results.
PMID 23690084