EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Discovery of 7-oxopyrazolo[1,5-a]pyrimidine-6-carboxamides as potent and selective CB(2) cannabinoid receptor inverse agonists.
PMID 23697626
Abstract
We recently described the medicinal chemistry of a new series of heteroaryl-4-oxopyridine/7-oxopyrimidines as CB2 receptor partial agonists, showing that the functionality of these ligands is controlled by the nature of the heteroaryl function condensed with the pyridine ring. We describe herein the design and synthesis of the 7-oxopyrazolo[1,5-a]pyrimidine-6-carboxamides, structural isomers of our previously reported pyrazolo[3,4-b]pyridines. All of the new compounds showed high affinity and selectivity for the CB2 receptor in the nanomolar range. In 3,5-cyclic adenosine monophosphate (cAMP) assays, the novel series shows stimulatory effects on forskolin-induced cAMP production acting as inverse agonists.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A6885
Adenosine 3′,5′-cyclic monophosphate sodium salt monohydrate, ≥98.0% (HPLC), powder
C10H11N5NaO6P · H2O