EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
The role of focal adhesion kinase in BMP4 induction of mesenchymal stem cell adipogenesis.
PMID 23702483
Abstract
Obesity is characterized by excessive adipocytic number growth and resultant adipose tissue hyperplasia. However, molecular mechanisms of abnormal recruitment of new adipocytes from precursor cells are not fully known. Several studies showed that bone morphogenetic proteins (BMPs) also play a role in inducing mesenchymal stem cells (MSCs) to commit to adipocytes. We tested the hypothesis that focal adhesion kinase (FAK), one of the vital focal adhesion signaling molecules, is required for BMP4 induction of MSC adipogenesis. BMP4 exposure triggered FAK activation at pY397 auto-phosphorylation site in murine C3H10T1/2 MSCs. Interestingly, silencing FAK by small hairpin RNA (shRNA) significantly suppressed BMP4 induction of MSC adipogenic activities, including lipid accumulation and expression of key adipogenic genes (C/EBPα, PPARγ, aP2), as relative to shRNA vector control. As a potential molecular mechanism, BMP4-triggered phosphorylation in Smad1/5/8 and p38 was significantly downregulated by shRNA-FAK. Pharmacological FAK inhibitor 14 provided similar results in BMP4-mediated MSC adipogenesis and Smad/p38 signaling. Our data clearly suggest a link between FAK and BMP4 induction of MSC adipogenesis, and may indicate a potential therapeutic approach targeting FAK for dealing with obesity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0837 FAK Inhibitor 14, ≥95% (HPLC)
C6H10N4 · 4HCl