EMAIL THIS PAGE TO A FRIEND
eLife
Structure of a pore-blocking toxin in complex with a eukaryotic voltage-dependent K(+) channel.
PMID 23705070
Abstract
Pore-blocking toxins inhibit voltage-dependent K(+) channels (Kv channels) by plugging the ion-conduction pathway. We have solved the crystal structure of paddle chimera, a Kv channel in complex with charybdotoxin (CTX), a pore-blocking toxin. The toxin binds to the extracellular pore entryway without producing discernable alteration of the selectivity filter structure and is oriented to project its Lys27 into the pore. The most extracellular K(+) binding site (S1) is devoid of K(+) electron-density when wild-type CTX is bound, but K(+) density is present to some extent in a Lys27Met mutant. In crystals with Cs(+) replacing K(+), S1 electron-density is present even in the presence of Lys27, a finding compatible with the differential effects of Cs(+) vs K(+) on CTX affinity for the channel. Together, these results show that CTX binds to a K(+) channel in a lock and key manner and interacts directly with conducting ions inside the selectivity filter. DOI:http://dx.doi.org/10.7554/eLife.00594.001.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C7802 Charybdotoxin, ≥90% (HPLC)
C176H277N57O55S7
47431
Fmoc-Phe(4-I)-OH, ≥97.0%
C24H20INO4