EMAIL THIS PAGE TO A FRIEND
The American journal of emergency medicine
Cardiac anaphylaxis: a case of acute ST-segment elevation myocardial infarction after IM epinephrine for anaphylactic shock.
PMID 23706578
Abstract
A 62-year-old male smoker with no other comorbidities presented to emergency department with systemic anaphylaxis, due to oral diclofenac for toothache. He developed acute anterior wall myocardial infarction following IM epinephrine 1 mg 1:1000. Primary percutaneous coronary intervention was done, which showed a thrombus in the mid left anterior descending artery with no evidence of obstructive coronary artery disease after thrombus aspiration.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D1039990 Diclofenac potassium, European Pharmacopoeia (EP) Reference Standard
C14H10Cl2KNO2