EMAIL THIS PAGE TO A FRIEND
Plant foods for human nutrition (Dordrecht, Netherlands)
Yacon (Smallanthus sonchifolius): a functional food.
PMID 23709016
Abstract
Yacon (Smallanthus sonchifolius) is an Andean tuberous root that is regarded as a functional food given that it contains fructooligosaccharides (FOS), inulin and phenolic compounds. The consumption of FOS and inulin improves the growth of bifidobacteria in the colon, enhances mineral absorption and gastrointestinal metabolism and plays a role in the regulation of serum cholesterol. Furthermore, the literature reports that the consumption of these prebiotics promotes a positive modulation of the immune system, improving resistance to infections and allergic reactions. Certain studies have demonstrated the potential of yacon as an alternative food source for those patients with conditions that require dietary changes. This review intends to describe the potential of yacon as a prebiotic and its cultivation and industrial processing for human consumption.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I2255
Inulin from chicory
I3754
Inulin from dahlia tubers