EMAIL THIS PAGE TO A FRIEND
Digestive diseases and sciences
NOD2 mutations affect muramyl dipeptide stimulation of human B lymphocytes and interact with other IBD-associated genes.
PMID 23709157
Abstract
Genetic and functional studies have associated variants in the NOD2/CARD15 gene with Crohn's disease. This study aims to replicate the association of three common NOD2 mutations with Crohn's disease, study its effect on NOD2 expression in B cells and its interaction with other IBD-associated genes. A total of 294 IBD patients (179 familial IBD, 115 sporadic IBD) and 298 unrelated healthy controls were from central Pennsylvania. NOD2 mutations were analyzed by primer-specific amplification, PCR based-RFLP, and validated with the ABI SNPlexM genotyping system. Gene-gene interaction was studied using a statistical model for epistasis analysis. Three common NOD2 mutations are associated with Crohn's disease (p=5.08×10(-7), 1.67×10(-6), and 1.87×10(-2) for 1007fs, R720W, and G908R, respectively), but not with ulcerative colitis (p=0.1046, 0.1269, and 0.8929, respectively). For IBD overall, 1007finsC (p=4.4×10(-5)) and R720W (p=9.24×10(-5)) were associated with IBD, but not G908R (p=0.1198). We revealed significant interactions of NOD2 with other IBD susceptibility genes IL23R, DLG5, and OCTN1. We discovered that NOD2 was expressed in both normal human peripheral blood B cells and in EBV-transformed B cell lines. Moreover, we further demonstrated that muramyl dipeptide (MDP) stimulation of B lymphocytes up-regulated expression of NF-κB-p50 mRNA. NOD2 is expressed in peripheral B cells, and the up-regulation of NOD2 expression by MDP was significantly impaired by NOD2 mutations. The finding suggests a possible role of NOD2 in the immunological response in IBD pathogenesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A9519
N-Acetylmuramyl-L-alanyl-D-isoglutamine hydrate, ≥98% (TLC)
C19H32N4O11 · xH2O