EMAIL THIS PAGE TO A FRIEND
Clinical radiology
Re: Comprehensive MRA of the lower limbs including high-resolution extended-phase infra-inguinal imaging with gadobenate dimeglumine: initial experience with inter-individual comparison to the blood-pool contrast agent gadofosveset trisodium. A reply.
PMID 23711302