EMAIL THIS PAGE TO A FRIEND
International journal of cancer
Influence of soluble or matrix-bound isoforms of vascular endothelial growth factor-A on tumor response to vascular-targeted strategies.
PMID 23712501
Abstract
Antiangiogenic therapy based on blocking the actions of vascular endothelial growth factor-A (VEGF) can lead to "normalization" of blood vessels in both animal and human tumors. Differential expression of VEGF isoforms affects tumor vascular maturity, which could influence the normalization process and response to subsequent treatment. Fibrosarcoma cells expressing only VEGF120 or VEGF188 isoforms were implanted either subcutaneously (s.c.) or in dorsal skin-fold "window" chambers in SCID mice. VEGF120 was associated with vascular fragility and hemorrhage. Tumor-bearing mice were treated with repeat doses of SU5416, an indolinone receptor tyrosine kinase inhibitor with activity against VEGFR-2 and proven preclinical ability to induce tumor vascular normalization. SU5416 reduced vascularization in s.c. implants of both VEGF120 and VEGF188 tumors. However, in the window chamber, SU5416 treatment increased red cell velocity in VEGF120 (representing vascular normalization) but not VEGF188 tumors. SU5416 treatment had no effect on growth or necrosis levels in either tumor type but tended to counteract the increase in interstitial fluid pressure seen with growth of VEGF120 tumors. SU5416 pretreatment resulted in the normally fragile blood vessels in VEGF120-expressing tumors becoming resistant to the vascular damaging effects of the tubulin-binding vascular disrupting agent (VDA), combretastatin A4 3-O-phosphate (CA4P). Thus, vascular normalization induced by antiangiogenic treatment can reduce the efficacy of subsequent VDA treatment. Expression of VEGF120 made tumors particularly susceptible to vascular normalization by SU5416, which in turn made them resistant to CA4P. Therefore, VEGF isoform expression may be useful for predicting response to both antiangiogenic and vascular-disrupting therapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML1131
Fosbretabulin disodium, ≥98% (HPLC)
C18H19O8P · 2Na