EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica
[Protective effects of polysaccharides from Dendrobium huoshanense on CCl4-induced acute liver injury in mice].
PMID 23713278
Abstract
To study the protective effects of polysaccharides from Dendrobium huoshanense (DHP) against CCl4-induced liver injury in mice. Eighty male Kunming mice were randomly divided into normal control group, model control group, dextran control group, starch control group, hydrolyzate control group, three different dose of DPH groups consisting of high-dosage group, middle-dosage group and low-dosage group (200, 100, 50 mg x kg(-1)). Each group contained ten mice. The mice were treated with DHP via intragastric administration for 15 days before treatment of 50% CCl4 in olive oil for consecutive two days. Both alanine aminotransferase (ALT) and aspartate aminotransferase (AST) activities in serum and superoxide dismutase (SOD) activities and malondialdehyde (MDA) contents in liver tissues were determined in all groups. Immunohistochemistry was used to detect the expression of TNF-alpha in hepatic tissue. Hepatic histopathological examination was observed. DHP effectively decreased the activities of ALT and AST in serum and the contents of hepatic MDA, and restored hepatic SOD activities in acute liver injury mice. Liver tissue damage induced by CCl4 was ameliorated in mice with DHP administration through histopathology examination. Furthermore, the expression of TNF-alpha was greatly decreased in groups treated with polysaccharides. DHP has a significantly hepatoprotective effect on CCl4-induced acute liver injury in mice. Protective effect of DHP on the liver may be related to its function of scavenging free radicals and inhibiting lipid peroxidation and TNF-alpha expression.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
270652
Carbon tetrachloride, for HPLC, ≥99.9%
CCl4
319961
Carbon tetrachloride, reagent grade, 99.9%
CCl4
48604 Carbon tetrachloride solution, certified reference material, 200 μg/mL in methanol
G2751 Glutamic-Oxalacetic Transaminase from porcine heart, Type I, ammonium sulfate suspension, 200-500 units/mg protein
G8255 Glutamic-Pyruvic Transaminase from porcine heart, lyophilized powder, ≥75 units/mg protein
G9880 Glutamic-Pyruvic Transaminase from porcine heart, ammonium sulfate suspension, ≥75 units/mg protein
S9697
Superoxide Dismutase bovine, recombinant, expressed in E. coli, lyophilized powder, ≥2500 units/mg protein, ≥90% (SDS-PAGE)
S5395
Superoxide Dismutase from bovine erythrocytes, BioReagent, ≥3,000 units/mg protein, suitable for cell culture, lyophilized powder
S7571
Superoxide Dismutase from bovine erythrocytes, lyophilized powder, ≥3,000 units/mg protein
S7446
Superoxide Dismutase from bovine erythrocytes, BioUltra, lyophilized powder, ≥4,500 units/mg protein, ≥97% (SDS-PAGE)
S8160
Superoxide Dismutase from bovine liver, lyophilized powder, ≥1500 units/mg protein
S8409
Superoxide Dismutase from bovine liver, ammonium sulfate suspension, 2,000-6,000 units/mg protein (biuret)
S5639
Superoxide Dismutase from Escherichia coli, lyophilized powder, ≥1,000 units/mg protein
S5389
Superoxide Dismutase from Escherichia coli, lyophilized powder, ≥1,000 units/mg protein
S9636
Superoxide Dismutase from human erythrocytes, essentially salt-free, lyophilized powder, ≥2,500 units/mg protein
S9076 Superoxide Dismutase I human, recombinant, expressed in E. coli