EMAIL THIS PAGE TO A FRIEND
Fiziolohichnyi zhurnal (Kiev, Ukraine : 1994)
[Hydrogen sulfide inhibits Ca(2+)-induced mitochondrial permeability transition pore opening in spontaneously hypertensive rats].
PMID 23713344
Abstract
In experiments in vivo and in vitro on the mitochondria isolated from the control and spontaneously hypertensive rats (SHR) hearts, we studied the effects of a donor of hydrogen sulfide (H2S), NaHS, and H2S biosynthesis substrate, L-cysteine, on the sensitivity of the mitochondrial permeability transition pore (mPTP) opening to its natural inductor, Ca2+. We found that NaHS (10(-4), 10(-5) and 5 10(-5) mol/l) influenced the mitochondrial swelling in a concentration-dependent manner in control and spontaneously hypertensive rats. The H2S donor NaHS used in physiological concentrations (10(-6), 10(-5) and 5 10(-5) mol/l) exerted the inhibiting effect on the Ca(2+)-induced mPTP opening in control hearts (corresponding values of such effect were 31, 76, and 100%, respectively), while in spontaneously hypertensive rats hearts the protector effect of NaHS was observed only at its concentration of 10(-5) - 10(-4) mol/l. In experiments in vivo, single intraperitoneal injections of L-cysteine (10(-3) mol/kg) resulted in a decrease in the sensitivity of mPTP to it's inductor Ca2+ in control rats and SHR. In experiments in vivo in which we used a specific blocker of cystathionine-gamma-lyase, propargylglycine (10(-4) mol/kg), with the further injections of L-cysteine we observed a decrease in the threshold Ca2+ concentration (that induce the mitochondrial swelling) by three orders of magnitude in SHR, but in control rats did not effect of L-cysteine. Thus, both endogenous and exogenous hydrogen sulfide inhibits Ca(2+)-induced mitochondrial permeability transition pore opening, indicating its protective effect on pore formation in spontaneously hypertensive rats hearts. Therefore, our studies are indicative of the involvement of H2S in modulation of changes in the permeability of mitochondrial membranes, which can be an important regulatory factor in the development of cardiovascular diseases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C3290000
Cysteine hydrochloride monohydrate, European Pharmacopoeia (EP) Reference Standard
C3H7NO2S · HCl · H2O
G2879
Glycine hydrochloride, ≥99% (HPLC)
C2H5NO2 · HCl
55097
Glycine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
C2H5NO2 · HCl
81838
L-C-Propargylglycine, ≥99.0% (TLC)
C5H7NO2
14495 L-Cysteine 0.1 M solution
C7880
L-Cysteine hydrochloride monohydrate, reagent grade, ≥98% (TLC)
C3H7NO2S · HCl · H2O
C6852
L-Cysteine hydrochloride monohydrate, from non-animal source, cell culture tested, meets EP, USP testing specifications
C3H7NO2S · HCl · H2O
C5610
L-Cysteine hydrochloride monohydrate, PharmaGrade, Ajinomoto, EP, USP, manufactured under appropriate GMP controls for Pharma or Biopharmaceutical production, suitable for cell culture
C3H7NO2S · HCl · H2O
30122
L-Cysteine hydrochloride monohydrate, tested according to Ph.Eur.
C3H7NO2S · HCl · H2O
W779989
L-Cysteine hydrochloride monohydrate, FG, produced by Wacker Chemie AG, Burghausen, Germany, Food, 20 mesh
C3H7NO2S · HCl · H2O
PHR1102
L-Cysteine hydrochloride monohydrate, Pharmaceutical Secondary Standard; Certified Reference Material
C3H7NO2S · HCl · H2O
30129
L-Cysteine hydrochloride monohydrate, BioUltra, ≥99.0% (RT)
C3H7NO2S · HCl · H2O
778451
L-Cysteine hydrochloride monohydrate, Produced by Wacker Chemie AG, Burghausen, Germany, Life Science, 98.5-101.0% (Ph.Eur (2.2.20))
C3H7NO2S · HCl · H2O