EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
A model system for the synthesis of complanadine alkaloids by "diverted Kondrat'eva" oxazole-olefin cycloaddition.
PMID 23713565
Abstract
A synthetic approach to complanadine alkaloids is described which employs a Kondrat'eva reaction to construct the pyridine rings. The viability of this approach is demonstrated by its application to a model substrate accessed from unfunctionalized decalin. The key transformation affords the desired tetracyclic architecture with unprecedented incorporation of substituents on the pyridine ring, implicating the oxazole α-hydroxy group as an active participant in the cycloadduct fragmentation process.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
192201
2,4,6-Trimethylbenzenesulfonohydrazide, 97%
C9H14N2O2S