EMAIL THIS PAGE TO A FRIEND
Journal of natural products
Alkaloids and polyketides from Penicillium citrinum, an endophyte isolated from the Moroccan plant Ceratonia siliqua.
PMID 23713692
Abstract
The endophytic fungus Penicillium citrinum was isolated from a fresh stem of the Moroccan plant Ceratonia siliqua. Extracts of P. citrinum grown on rice and white bean media yielded five new compounds, namely, citriquinochroman (1), tanzawaic acids G and H (2 and 3), 6-methylcurvulinic acid (4), 8-methoxy-3,5-dimethylisoquinolin-6-ol (5), and one new natural product, 1,2,3,11b-tetrahydroquinolactacide (6), which had previously been described as a synthetic compound. In addition, 13 known compounds including seven alkaloids and six polyketides were isolated. The structures of the new compounds were unambiguously determined on the basis of one- and two-dimensional NMR spectroscopy as well as by high-resolution mass spectrometry. Citriquinochroman (1) features a new skeleton, consisting of quinolactacide and (3S)-6-hydroxy-8-methoxy-3,5-dimethylisochroman linked by a C-C bond. 1,2,3,11b-Tetrahydroquinolactacide (6) may be a biogenetic precursor of quinolactacide. Citriquinochroman (1) showed cytotoxicity against the murine lymphoma L5178Y cell line with an IC50 value of 6.1 μM, while the other compounds were inactive (IC50 >10 μM) in this assay.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
L5178Y, 87111908