EMAIL THIS PAGE TO A FRIEND
Drug Metabolism Reviews
Synthesis of fatty acid ethyl esters in mammalian tissues after ethanol exposure: a systematic review of the literature.
PMID 23713893
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
24102
Ethanol, absolute, ≥99.8% (GC)
C2H6O
24105
Ethanol, puriss., meets analytical specification of Ph. Eur., BP, 96.0-97.2%
C2H6O
24194
Ethanol, absolute, denaturated with 0.5-1.5 Vol.% 2-butanone and approx. 0.001% Bitrex (GC), ≥98% (GC)
C2H6O
E-034
Ethanol Calibration Kit, ampule of 10 × 1.2 mL, certified reference material
C2H6O
E-040
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material
C2H6O
E-031
Ethanol-100 (10 ampules/kit), 100 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material
C2H6O
E-038
Ethanol-100 (5 ampules/kit), 100 mg/dL in H2O, ampule of 5 × 5 mL, certified reference material
C2H6O
E-041
Ethanol-150, 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material
C2H6O
E-056
Ethanol-20 (10 ampules/kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material
C2H6O
E-043
Ethanol-20 (5 ampules/kit), 20 mg/dL in H2O, ampule of 5 × 5 mL, certified reference material
C2H6O
E-032
Ethanol-200 (10 ampules/kit), 200 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material
C2H6O
E-039
Ethanol-200 (5 ampules/kit), 200 mg/dL in H2O, ampule of 5 × 5 mL, certified reference material
C2H6O
E-035
Ethanol-25, 25 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material
C2H6O
E-033
Ethanol-300, 300 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material
C2H6O
E-045
Ethanol-40, 40 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material
C2H6O
E-036
Ethanol-400 (10 ampules/kit), 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material
C2H6O
E-044
Ethanol-400 (5 ampules/kit), 400 mg/dL in H2O, ampule of 5 × 5 mL, certified reference material
C2H6O
E-029
Ethanol-50, 50 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material
C2H6O
E-053
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material
C2H6O
E-030
Ethanol-80, 80 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material
C2H6O
E-037
Ethanol-80, 80 mg/dL in H2O, ampule of 5 × 5 mL, certified reference material
C2H6O
187380 Ethyl alcohol, denatured, reagent grade
459844
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
C2H6O
E7023
Ethyl alcohol, Pure, 200 proof, for molecular biology
C2H6O
493546
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
C2H6O
459828
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
C2H6O
E7148
Ethyl alcohol, Pure, 190 proof, for molecular biology
C2H6O
270741
Reagent Alcohol, for HPLC
245119
Reagent Alcohol, spectrophotometric grade
277649
Reagent Alcohol, anhydrous, ≤0.003% water