EMAIL THIS PAGE TO A FRIEND
Chemosphere
Biosorption of phenanthrene by pure algae and field-collected planktons and their fractions.
PMID 23714149
Abstract
The biosorption isotherms for phenanthrene (Phen) by cultured algae, field-collected plankton, and market algae samples (OSs) and their fractions (lipid-LP, lipid free carbon-LF, alkaline nonhydrolyzable carbon-ANHC, and acid nonhydrolyzable carbon-NHC) were established. All the biosorption isotherms are well fitted by the Freundlich model. The biosorption isotherms for the ANHC and NHC fractions are nonlinear and for the other fractions are linear. It was found that the NHC fractions are chemically and structurally different from other fractions by using elemental analysis and Fourier transformed infrared spectroscopy (FTIR), consisting mainly of aliphatic polymethylene carbon. The average KOC values for Phen at Ce=0.005Sw are 10706±2768mLg(-1) and 95843±55817mLg(-1) for the bulk market algal samples and their NHC isolates, respectively. As the NHC fraction for Porphyra contains higher polymethylene carbon than that for Seaweed or Spirulina, it exhibits higher biosorption capacity. Moreover, the logKOC values are significantly higher for the field-collected samples than for the market algae and cultured algae samples. The multivariate correlation shows that the logKOC values are positively related to the LP contents, and negatively to the C/N ratios for the original algal samples. Furthermore, the logKOC values are negatively related to the polarity indices (O/C and O+N/C) for the original samples and their fractions excluding LP fractions. These observations help to understand the role of polarity, LP and NHC fractions, and aliphatic structures in the biosorption of Phen, which requires more attention in the examination of sorption processes in the natural environment.