EMAIL THIS PAGE TO A FRIEND
BMC psychiatry
Tardive and spontaneous dyskinesia incidence in the general population.
PMID 23714238
Abstract
To identify the incidence rate of spontaneous dyskinesia (SD) and tardive dyskinesia (TD) in a general population and to examine the association between dykinesia and potential risk factors (exposure to metoclopramide [MCP], antipsychotic drugs, and history of diabetes and psychoses). A retrospective cohort study was conducted for the years 2001 through 2010, based on medical claims data from the Deseret Mutual Benefit Administrators (DMBA). Thirty-four cases of TD and 229 cases of SD were identified. The incidence rate of TD among persons previously prescribed an antipsychotic or metoclopramide (MCP) (per 1,000) was 4.6 (1.6-7.7) for those with antipsychotic drug use only, 8.5 (4.8-12.2) for those with MCP use only, and 15.0 (2.0-28.1) for those with both antipsychotic and MCP use. In the general population, the incidence rate (per 100,000 person-years) of TD was 4.3 and of probable SD was 28.7. The incidence rates of TD and SD increased with age and were greater for females. Those with diabetes or psychoses had almost a 3-fold greater risk of TD than those without either of these diseases. Persons with schizophrenia had 31.2 times increased risk of TD than those without the disease. Positive associations also existed between the selected diseases and the incidence rate of probable SD, with persons with schizophrenia having 4.4 times greater risk of SD than those without the disease. SD and TD are rare in this general population. Diabetes, psychoses, and especially schizophrenia are positively associated with SD and TD. A higher proportion of those with SD present with spasm of the eyelid muscles (blepharospasm) compared more with the TD cases who present more with orofacial muscular problems.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1440808 Metoclopramide hydrochloride, United States Pharmacopeia (USP) Reference Standard
C14H22ClN3O2 · HCl · H2O