EMAIL THIS PAGE TO A FRIEND
Journal of clinical oncology : official journal of the American Society of Clinical Oncology
Unusual case of recurrent extraneural metastatic medulloblastoma in a young adult: durable complete remission with Ewing sarcoma chemotherapy regimen and consolidation with autologous bone marrow transplantation and local radiation.
PMID 23715566