EMAIL THIS PAGE TO A FRIEND
Journal of clinical oncology : official journal of the American Society of Clinical Oncology
Histologic transformation of Epstein-Barr virus-positive diffuse large B-cell lymphoma of the elderly.
PMID 23715578