EMAIL THIS PAGE TO A FRIEND
Annals of the rheumatic diseases
A two-step treatment strategy trial in patients with early arthritis aimed at achieving remission: the IMPROVED study.
PMID 23716067
Abstract
To assess which treatment strategy is most effective in inducing remission in early (rheumatoid) arthritis. 610 patients with early rheumatoid arthritis (RA 2010 criteria) or undifferentiated arthritis (UA) started treatment with methotrexate (MTX) and a tapered high dose of prednisone. Patients in early remission (Disease Activity Score <1.6 after 4 months) tapered prednisone to zero and those with persistent remission after 8 months, tapered and stopped MTX. Patients not in early remission were randomised to receive either MTX plus hydroxychloroquine plus sulfasalazine plus low-dose prednisone (arm 1) or to MTX plus adalimumab (ADA) (arm 2). If remission was present after 8 months both arms tapered to MTX monotherapy; if not, arm 1 changed to MTX plus ADA and arm 2 increased the dose of ADA. Remission rates and functional and radiological outcomes were compared between arms and between patients with RA and those with UA. 375/610 (61%) patients achieved early remission. After 1 year 68% of those were in remission and 32% in drug-free remission. Of the randomised patients, 25% in arm 1 and 41% in arm 2 achieved remission at year 1 (p<0.01). Outcomes were comparable between patients with RA and those with UA. Initial MTX and prednisone resulted in early remission in 61% of patients with early (rheumatoid) arthritis. Of those, 68% were in remission and 32% were in drug-free remission after 1 year. In patients not in early remission, earlier introduction of ADA resulted in more remission at year 1 than first treating with disease-modifying antirheumatic drug combination therapy plus prednisone.