EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica
[Study on intervention effect of Danggui Shaoyao San on rats with cirrhotic ascites].
PMID 23717970
Abstract
To investigate the intervention effect of Danggui Shaoyao San on rats with cirrhotic ascites, and discuss the effect of arginine vasopressin (AVP) on cirrhotic ascites. Male SD rats were randomly divided into the control group, the model group, Danggui Shaoyao San low, middle and high dose groups. The cirrhotic ascites rat model was established by CCl4 combined with phenobarbital. Their urines were collected at 24 h to observe urine excretion of each group. Filter papers were used to determine the amount of ascites. The levels of serum alanine aminotransferasa (ALT) , aspartate aminotransferase (AST) were detected by the automatic biochemistry analyzer. Plasma prothrombin time (PT) was evaluated by the blood coagulation analyzer. The concentration of AVP in plasma was detected by enzyme-linked immunosorbent assay (ELISA). Pathological changes in livers were observed by HE staining. Compared with the model group, the Danggui Shaoyao San group showed significant improvement in live indexes, with notable decrease in serum ALT and AST and the time of PT, improvement in liver pathological changes. Simultaneously, the amount of ascites decreased to varying degrees, with notable increase in urine in 24 h and decrease in AVP concentration in plasma. Danggui Shaoyao San can notably improve liver functions of rats with cirrhotic ascites, reduce the generation of ascites and delay the progress of liver pathological changes. Its mechanism may be related to AVP.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
V0377
[Arg8]-Vasopressin solution, Grade VI (synthetic), ~100 I.U./mL in 0.9% NaCl
C46H65N15O12S2