EMAIL THIS PAGE TO A FRIEND
Klinichna khirurhiia
[Epidural pharmacotherapy in the treatment of the pain syndrome in osteochondrosis complicated by intervertebral discs protrusion of lumbar part of the vertebral column].
PMID 23718034
Abstract
Experience of application of intralaminar epidural pharmacotherapy for the treatment of the pain syndrome in the patients, suffering osteochondrosis of lumbar portion of vertebral column, complicated by intervertebral discs protrusion, was summarized. The method introduction permits to escape the operative intervention performance for complicated osteochondrosis.