EMAIL THIS PAGE TO A FRIEND
Mikrobiologiia
[Comparison of the adaptation potential of Arthrobacter oxydans and Acinetobacter lwoffii isolated from permafrost sediments with their collection analogues].
PMID 23718046