EMAIL THIS PAGE TO A FRIEND
Mikrobiolohichnyi zhurnal (Kiev, Ukraine : 1993)
[Influence of transition metal compounds on superoxide dismutase activity of sulfur reducing Desulfuromonas acetoxidans bacteria].
PMID 23720962
Abstract
Superoxide dismutase, as one of the enzymes of cells' antioxidant defensive system, catalyzes superoxide anion-radical (O2-) dismutation with O2 and H2O2 forming. The influence of such transition metal compounds, as FeSO4, FeCl3, MnCl2, NiCl2, and CoCl2 on superoxide dismutase activity of sulfur-reducing Desulfuromonas acetoxidans bacteria has been investigated. Maximal activity of the investigated enzyme has been observed accordingly under the influence of 1.0 mM of NiCl2, 2.0 mM of CoCl2 and MnCl2 on the second day and under the influence of 1.0 mM of FeCl3 and FeSO4 respectively, on the third day of growth in comparison with control samples. An increase of incubation time and concentration of metal compound in the medium caused the inhibition of superoxide dismutase activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
356891
Cobalt, foil, thickness 1.0 mm, 99.95% trace metals basis
Co
266671
Cobalt, foil, thickness 0.25 mm, ≥99.99% trace metals basis
Co
326380
Cobalt, foil, thickness 0.1 mm, ≥99.99%
Co
356867
Cobalt, foil, thickness 0.1 mm, 99.95% trace metals basis
Co
203076
Cobalt, granular, 99.99% trace metals basis
Co
266655
Cobalt, pieces, 99.5% trace metals basis
Co
266647
Cobalt, powder, <150 μm, ≥99.9% trace metals basis
Co
266639
Cobalt, powder, 2 μm particle size, 99.8% trace metals basis
Co
398810
Cobalt, rod, diam. 5.0 mm, 99.95% trace metals basis
Co
266701
Cobalt, wire, diam. 1.0 mm, 99.995% trace metals basis
Co
697745
Cobalt, Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%
Co
15862
Cobalt chloride 0.1 M solution
60818
Cobalt(II) chloride, purum p.a., anhydrous, ≥98.0% (KT)
Cl2Co
232696
Cobalt(II) chloride, 97%
Cl2Co
F8263
Iron(II) sulfate heptahydrate, suitable for plant cell culture, ≥99%
FeO4S · 7H2O
F8633
Iron(II) sulfate heptahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
FeO4S · 7H2O
215422
Iron(II) sulfate heptahydrate, ACS reagent, ≥99.0%
FeO4S · 7H2O
F7002
Iron(II) sulfate heptahydrate, ReagentPlus®, ≥99%
FeO4S · 7H2O
12354
Iron(II) sulfate heptahydrate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.5-104.5% (manganometric)
FeO4S · 7H2O
236489
Iron(III) chloride hexahydrate, ACS reagent, 97%
Cl3Fe · 6H2O
F2877
Iron(III) chloride hexahydrate, reagent grade, ≥98%, chunks
Cl3Fe · 6H2O
44944
Iron(III) chloride hexahydrate, puriss. p.a., ACS reagent, crystallized, 98.0-102% (RT)
Cl3Fe · 6H2O
31232
Iron(III) chloride hexahydrate, puriss. p.a., Reag. Ph. Eur., ≥99%
Cl3Fe · 6H2O
12322
Iron(III) chloride solution, purum, 45% FeCl3 basis
Cl3Fe
450995
Manganese(II) chloride, anhydrous, beads, −10 mesh, 99.999% trace metals basis
Cl2Mn
429449
Manganese(II) chloride, anhydrous, beads, −10 mesh, 99.99% trace metals basis
Cl2Mn
328146
Manganese(II) chloride, beads, 98%
Cl2Mn
416479
Manganese(II) chloride, flakes, 97%
Cl2Mn
244589
Manganese(II) chloride, powder and chunks, ≥99% trace metals basis
Cl2Mn
48795
Manganese(II) chloride 0.1 M solution
Cl2Mn