EMAIL THIS PAGE TO A FRIEND
Nature methods
CLARITY for mapping the nervous system.
PMID 23722210
Abstract
With potential relevance for brain-mapping work, hydrogel-based structures can now be built from within biological tissue to allow subsequent removal of lipids without mechanical disassembly of the tissue. This process creates a tissue-hydrogel hybrid that is physically stable, that preserves fine structure, proteins and nucleic acids, and that is permeable to both visible-spectrum photons and exogenous macromolecules. Here we highlight relevant challenges and opportunities of this approach, especially with regard to integration with complementary methodologies for brain-mapping studies.