EMAIL THIS PAGE TO A FRIEND
Inflammation research : official journal of the European Histamine Research Society ... [et al.]
Blockade of cannabinoid receptors reduces inflammation, leukocyte accumulation and neovascularization in a model of sponge-induced inflammatory angiogenesis.
PMID 23722450
Abstract
Angiogenesis depends on a complex interaction between cellular networks and mediators. The endocannabinoid system and its receptors have been shown to play a role in models of inflammation. Here, we investigated whether blockade of cannabinoid receptors may interfere with inflammatory angiogenesis. Polyester-polyurethane sponges were implanted in C57Bl/6j mice. Animals received doses (3 and 10 mg/kg/daily, s.c.) of the cannabinoid receptor antagonists SR141716A (CB1) or SR144528 (CB2). Implants were collected at days 7 and 14 for cytokines, hemoglobin, myeloperoxidase, and N-acetylglucosaminidase measurements, as indices of inflammation, angiogenesis, neutrophil and macrophage accumulation, respectively. Histological and morphometric analysis were also performed. Cannabinoid receptors expression in implants was detected from day 4 after implantation. Treatment with CB1 or CB2 receptor antagonists reduced cellular influx into sponges at days 7 and 14 after implantation, although CB1 receptor antagonist were more effective at blocking leukocyte accumulation. There was a reduction in TNF-α, VEGF, CXCL1/KC, CCL2/JE, and CCL3/MIP-1α levels, with increase in CCL5/RANTES. Both treatments reduced neovascularization. Dual blockade of cannabinoid receptors resulted in maximum inhibition of inflammatory angiogenesis. Blockade of cannabinoid receptors reduced leukocyte accumulation, inflammation and neovascularization, suggesting an important role of endocannabinoids in sponge-induced inflammatory angiogenesis both via CB1 and CB2 receptors.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0800 Rimonabant hydrochloride, ≥98% (HPLC)
C22H21Cl3N4O · HCl