EMAIL THIS PAGE TO A FRIEND
Journal of leukocyte biology
Editorial: A missing link? Monocyte activation by uremic toxins in cardiorenal syndrome.
PMID 23723377
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I3875
Indoxyl sulfate potassium salt
C8H6KNO4S